2-(Aminomethyl)pyrazine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Pyrazin-2-ylmethanamine
OR40048-1G
OR40048-5G
OR40048-10G
APOSOR40048-1GEA
622
CZK
APOSOR40048-1G
APOSOR40048-5G
APOSOR40048-10G
2-(Aminomethyl)pyrazine 97%
2-(Aminomethyl)pyrazine
Vzorec:
C₅H₇N₃ MW: 109,1292 g/mol Bod varu: 87…88 °C/3mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD00673149 CAS číslo: 20010-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...