2-Chloro-6-methoxyphenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR400479-1G
OR400479-25G
OR400479-5G
APOSOR400479-1GEA
622
CZK
APOSOR400479-1G
APOSOR400479-25G
APOSOR400479-5G
2-Chloro-6-methoxyphenylboronic acid
2-Chloro-6-methoxyphenylboronic acid
Vzorec:
C₇H₈BClO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD04039878 CAS číslo: 385370-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...