2-Bromo-3-iodopyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR400475-1G
OR400475-25G
OR400475-5G
APOSOR400475-1GEA
662
CZK
APOSOR400475-1G
APOSOR400475-25G
APOSOR400475-5G
2-Bromo-3-iodopyridine
2-Bromo-3-iodopyridine
Vzorec:
C₅H₃BrIN Bod tání: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD09056782 CAS číslo: 265981-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...