5-Chlorobenzoxazole
Dodavatel: Apollo Scientific
OR400452-25G
OR400452-5G
APOSOR400452-25GEA
2610
CZK
APOSOR400452-25G
APOSOR400452-5G
5-Chlorobenzoxazole
5-Chlorobenzoxazole
Vzorec:
C₇H₄ClNO MW: 153.57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00216939 CAS číslo: 17200-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...