4-Iodo-3-nitroanisole
Dodavatel: Apollo Scientific
Synonyms:
1-Iodo-4-methoxy-2-nitrobenzene
OR400414-25G
OR400414-5G
APOSOR400414-25GEA
2440
CZK
APOSOR400414-25G
APOSOR400414-5G
4-Iodo-3-nitroanisole
4-Iodo-3-nitroanisole
Vzorec:
C₇H₆INO₃ MW: 279.03 Storage Temperature: Ambient |
MDL Number:
MFCD00024195 CAS číslo: 58755-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...