Tisk…

3-(Boc-aminomethyl)azetidine hydrochloride 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-butyl N-(azetidin-3-ylmethyl)carbamate

OR40040-1G OR40040-5G
APOSOR40040-1GEA 2150 CZK
APOSOR40040-1G APOSOR40040-5G
3-(Boc-aminomethyl)azetidine hydrochloride 96%
3-(Boc-aminomethyl)azetidine hydrochloride
Vzorec: C₉H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01861760
CAS číslo: 91188-15-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR