Tisk…

Methyl-2-chloro-5-iodobenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-chloro-5-iodobenzoate

OR400205-1G OR400205-5G OR400205-25G
APOSOR400205-1GEA 622 CZK
APOSOR400205-1G APOSOR400205-5G APOSOR400205-25G
Methyl-2-chloro-5-iodobenzoate
Methyl-2-chloro-5-iodobenzoate
Vzorec: C₈H₆ClIO₂
MW: 296,4900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779253
CAS číslo: 620621-48-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR