Tisk…

2-Bromo-3-chlorobenzaldehyde

Dodavatel: Apollo Scientific

OR400100-5G
APOSOR400100-5GEA 2530 CZK
APOSOR400100-5G
2-Bromo-3-chlorobenzaldehyde
2-Bromo-3-chlorobenzaldehyde
Vzorec: C₇H₄BrClO
MW: 219.4600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699536
CAS číslo: 74073-40-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR