Methyl-4-bromosalicylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 4-bromo-2-hydroxybenzoate
OR400066-1G
OR400066-5G
OR400066-25G
APOSOR400066-1GEA
622
CZK
APOSOR400066-1G
APOSOR400066-5G
APOSOR400066-25G
Methyl-4-bromosalicylate
Methyl-4-bromosalicylate
Vzorec:
C₈H₇BrO₃ MW: 231,0434 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07780736 CAS číslo: 22717-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...