2-Methoxy-5-nitrobenzonitrile
Dodavatel: Apollo Scientific
OR400055-1G
OR400055-5G
APOSOR400055-1GEA
1990
CZK
APOSOR400055-1G
APOSOR400055-5G
2-Methoxy-5-nitrobenzonitrile
2-Methoxy-5-nitrobenzonitrile
Vzorec:
C₈H₆N₂O₃ MW: 178,1448 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00035891 CAS číslo: 10496-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...