4-Chloroisatoic anhydride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione
OR400016-25G
APOSOR400016-25GEA
3410
CZK
APOSOR400016-25G
4-Chloroisatoic anhydride
4-Chloroisatoic anhydride
Vzorec:
C₈H₄ClNO₃ MW: 197,58 g/mol Bod tání: 255…259 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090431 CAS číslo: 40928-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...