Tisk…

3-Chloroisatoic anhydride

Dodavatel: Apollo Scientific
Varování

OR400010-1G OR400010-5G OR400010-25G
APOSOR400010-1GEA 1370 CZK
APOSOR400010-1G APOSOR400010-5G APOSOR400010-25G
3-Chloroisatoic anhydride
3-Chloroisatoic anhydride
Vzorec: C₈H₄ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD06809785
CAS číslo: 63497-60-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR