Tisk…

2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-(1,3,2-dioxaborinan-2-yl)benzoate

OR3999-1G OR3999-5G
APOSOR3999-1GEA 2240 CZK
APOSOR3999-1G APOSOR3999-5G
2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester 95%
2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester
Vzorec: C₁₂H₁₅BO₄
MW: 234,0561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659932
CAS číslo: 850567-60-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR