3-Bromo-5-methoxyphenol 97%
Dodavatel: Apollo Scientific
Synonyms:
3-Brom-5-hydroxyanisole
OR399011-250MG
OR399011-1G
OR399011-5G
APOSOR399011-250MGEA
1620
CZK
APOSOR399011-250MG
APOSOR399011-1G
APOSOR399011-5G
3-Bromo-5-methoxyphenol 97%
3-Bromo-5-methoxyphenol
Vzorec:
C₇H₇BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12405420 CAS číslo: 855400-66-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...