Tisk…

(2-Nitrophenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2-Nitrophenyl)acetic acid

OR3988-100G OR3988-25G
APOSOR3988-100GEA 1370 CZK
APOSOR3988-100G APOSOR3988-25G
(2-Nitrophenyl)acetic acid
(2-Nitrophenyl)acetic acid
Vzorec: (C₆H₄NO₂)CH₂COOH
MW: 181.15
Bod tání: 137…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007190
CAS číslo: 3740-52-1
EINECS: 223-128-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR