2-Chlor-4-methylbenzen-1-sulfochlorid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR3971-100G
OR3971-25G
OR3971-5G
APOSOR3971-100GEA
12570
CZK
APOSOR3971-100G
APOSOR3971-25G
APOSOR3971-5G
2-Chlor-4-methylbenzen-1-sulfochlorid 98%
2-Chlor-4-methylbenzen-1-sulfochlorid
Vzorec:
C₇H₆Cl₂O₂S Bod varu: 94…96 °C/0,05mmHg Bod tání: 34…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00203987 CAS číslo: 42413-03-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...