Tisk…

[4-(1-Cyano-1-methylethyl)phenyl]boronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(2-Cyanoprop-2-yl)benzeneboronic acid, 2-(4-Boronophenyl)-2-methylpropanenitrile

OR3950-100MG OR3950-5G
APOSOR3950-100MGEA 2070 CZK
APOSOR3950-100MG APOSOR3950-5G
[4-(1-Cyano-1-methylethyl)phenyl]boronic acid 98%
[4-(1-Cyano-1-methylethyl)phenyl]boronic acid
Vzorec: C₁₀H₁₂BNO₂
MW: 189,02 g/mol
Bod tání: 146…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659832
CAS číslo: 850568-67-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR