Tisk…

5-Chlorobenzofurazan-4-sulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Chloro-2,1,3-benzoxadiazole-4-sulphonyl chloride

OR3923-1G OR3923-250MG
APOSOR3923-1GEA 2480 CZK
APOSOR3923-1G APOSOR3923-250MG
5-Chlorobenzofurazan-4-sulphonyl chloride 97%
5-Chlorobenzofurazan-4-sulphonyl chloride
Vzorec: C₆H₂Cl₂N₂O₃S
MW: 253,06 g/mol
Bod tání: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00203878
CAS číslo: 175203-78-8
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR