Chloroacetamidine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloroacetimidamide hydrochloride, 2-Chloroacetamidine hydrochloride
OR3921-25G
OR3921-5G
APOSOR3921-25GEA
4300
CZK
APOSOR3921-25G
APOSOR3921-5G
Chloroacetamidine hydrochloride
Chloroacetamidine hydrochloride
Vzorec:
C₂H₆Cl₂N₂ MW: 128.99 Storage Temperature: Ambient |
MDL Number:
MFCD00053013 CAS číslo: 10300-69-3 EINECS: 233-675-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...