Tisk…

4-Chloromethyl-2-methylthiazole hydrochloride

Dodavatel: Apollo Scientific
Nebezpečí

OR3920-1G OR3920-5G
APOSOR3920-1GEA 622 CZK
APOSOR3920-1G APOSOR3920-5G
4-Chloromethyl-2-methylthiazole hydrochloride
4-Chloromethyl-2-methylthiazole hydrochloride
Vzorec: C₅H₇Cl₂NS
MW: 184,078 g/mol
Bod tání: 166…168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067725
CAS číslo: 77470-53-2
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR