{4-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR3842-1G
OR3842-500MG
APOSOR3842-1GEA
9920
CZK
APOSOR3842-1G
APOSOR3842-500MG
{4-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine 97%
{4-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine
Vzorec:
C₁₃H₂₁N₃ MW: 219.33 g/mol Hustota: 1.015 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD08060530 CAS číslo: 515162-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...