N-Methyl-4-nitroaniline 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR3796-25G
OR3796-500G
APOSOR3796-25GEA
0
CZK
APOSOR3796-25G
APOSOR3796-500G
N-Methyl-4-nitroaniline 97%
N-Methyl-4-nitroaniline
Vzorec:
C₇H₈N₂O₂ MW: 152.15 Bod varu: 291 Bod tání: 149…154 °C Hustota: 1.201 Storage Temperature: Ambient |
MDL Number:
MFCD00007305 CAS číslo: 100-15-2 EINECS: 202-823-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...