Tisk…

Methyl-5-bromo-2-hydroxyisonicotinate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 5-bromo-2-hydroxyisonicotinate

OR3795-1G OR3795-5G
APOSOR3795-1GEA 2980 CZK
APOSOR3795-1G APOSOR3795-5G
Methyl-5-bromo-2-hydroxyisonicotinate 98%
Methyl-5-bromo-2-hydroxyisonicotinate
Vzorec: C₇H₆BrNO₃
MW: 232,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027265
CAS číslo: 913836-17-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR