Tisk…

4-Bromobenzo[b]thiophene

Dodavatel: Apollo Scientific
Varování

OR3759-100G OR3759-25G
APOSOR3759-100GEA 14490 CZK
APOSOR3759-100G APOSOR3759-25G
4-Bromobenzo[b]thiophene
4-Bromobenzo[b]thiophene
Vzorec: C₈H₅BrS
MW: 213,0900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669544
CAS číslo: 5118-13-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR