Tisk…

2-Bromo-N,N-dimethylbenzamide 98%

Dodavatel: Apollo Scientific
Varování

OR3747-25G OR3747-5G
APOSOR3747-25GEA 6750 CZK
APOSOR3747-25G APOSOR3747-5G
2-Bromo-N,N-dimethylbenzamide 98%
2-Bromo-N,N-dimethylbenzamide
Vzorec: C₉H₁₀BrNO
MW: 228,0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463635
CAS číslo: 54616-47-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR