Tisk…

2-Quinoxalinecarboxylic acid

Dodavatel: Apollo Scientific
Varování

OR3738-10G OR3738-5G OR3738-25G
APOSOR3738-10GEA 1160 CZK
APOSOR3738-10G APOSOR3738-5G APOSOR3738-25G
2-Quinoxalinecarboxylic acid
2-Quinoxalinecarboxylic acid
Vzorec: C₉H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00012334
CAS číslo: 879-65-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR