5-Aminopicolinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid
OR3731-1G
OR3731-5G
OR3731-10G
APOSOR3731-1GEA
622
CZK
APOSOR3731-1G
APOSOR3731-5G
APOSOR3731-10G
5-Aminopicolinic acid
5-Aminopicolinic acid
Vzorec:
C₆H₆N₂O₂ MW: 138,13 g/mol Bod tání: 216…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684600 CAS číslo: 24242-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...