Tisk…

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 5,6-Diamino-1,3-benzodioxole dihydrochloride, 1,3-Benzodioxole-5,6-diamine dihydrochloride, 1,2-Diamino-4,5-methylenedioxybenzene dihydrochloride, MDB, Benzo[1,3]dioxole-5-6-diamine dihydrochloride

OR3723-1G OR3723-250MG OR3723-50MG
APOSOR3723-1GEA 12300 CZK
APOSOR3723-1G APOSOR3723-250MG APOSOR3723-50MG
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride
Vzorec: C₇H₈N₂O₂·2HCl
MW: 225,07 g/mol
Bod tání: 176…179 °C
Storage Temperature: Freezer
MDL Number: MFCD00037497
CAS číslo: 81864-15-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR