1-(4-Bromobenzyl)piperidine
Dodavatel: Apollo Scientific
|
Varování
|
OR3709-1G
OR3709-5G
APOSOR3709-1GEA
0
CZK
APOSOR3709-1G
APOSOR3709-5G
1-(4-Bromobenzyl)piperidine
1-(4-Bromobenzyl)piperidine
Vzorec:
C₁₂H₁₆BrN Storage Temperature: Refrigerator |
MDL Number:
MFCD06080709 CAS číslo: 178162-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...