Tisk…

5-Bromo-2-chloro-4-(methylthio)pyrimidine 97%

Dodavatel: Apollo Scientific
Varování

OR3705-1G OR3705-5G
APOSOR3705-1GEA 1290 CZK
APOSOR3705-1G APOSOR3705-5G
5-Bromo-2-chloro-4-(methylthio)pyrimidine 97%
5-Bromo-2-chloro-4-(methylthio)pyrimidine
Vzorec: C₅H₄BrClN₂S
MW: 239,5200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781218
CAS číslo: 59549-51-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR