5-Bromo-2-chloroisonicotinic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR3702-100G
OR3702-25G
APOSOR3702-100GEA
6290
CZK
APOSOR3702-100G
APOSOR3702-25G
5-Bromo-2-chloroisonicotinic acid 97%
5-Bromo-2-chloroisonicotinic acid
Vzorec:
C₆H₃BrClNO₂ Bod tání: 220…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD07375114 CAS číslo: 886365-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...