Tisk…

4-Amino-N-(2-chlorophenyl)benzamide ≥98%

Dodavatel: Apollo Scientific

OR370048-25G OR370048-5G
APOSOR370048-25GEA 25820 CZK
APOSOR370048-25G APOSOR370048-5G
4-Amino-N-(2-chlorophenyl)benzamide ≥98%
4-Amino-N-(2-chlorophenyl)benzamide
Vzorec: C₁₃H₁₁ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00628973
CAS číslo: 888-79-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR