Tisk…

3-Hydroxyindole

Dodavatel: Apollo Scientific
Varování

OR3695-250MG OR3695-100MG
APOSOR3695-250MGEA 10350 CZK
APOSOR3695-250MG APOSOR3695-100MG
3-Hydroxyindole
3-Hydroxyindole
Vzorec: C₈H₇NO
MW: 133,15 g/mol
Storage Temperature: Freezer
CAS číslo: 480-93-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR