Tisk…

1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Pyrrolidine-3-carboxylic acid, N-BOC protected

OR3670-1G OR3670-5G
APOSOR3670-1GEA 622 CZK
APOSOR3670-1G APOSOR3670-5G
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
Vzorec: C₁₀H₁₇NO₄
MW: 215,25 g/mol
Bod tání: 133…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179181
CAS číslo: 59378-75-5
UN: 3077
ADR: 9+EHS,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR