4-Methoxyphenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Methoxybenzeneboronic acid, Dihydroxy(4-methoxyphenyl)borane, (4-Methoxyphenyl)metaboric acid
OR3665-100G
OR3665-25G
OR3665-500G
APOSOR3665-100GEA
1540
CZK
APOSOR3665-100G
APOSOR3665-25G
APOSOR3665-500G
4-Methoxyphenylboronic acid
4-Methoxyphenylboronic acid
Vzorec:
C₇H₉BO₃ MW: 151,96 g/mol Bod tání: 204…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039139 CAS číslo: 5720-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...