3-Methoxyphenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Methoxyphenyl)metaboric acid, Dihydroxy(3-methoxyphenyl)borane, 3-Methoxybenzeneboronic acid
OR3664-25G
OR3664-100G
APOSOR3664-25GEA
827
CZK
APOSOR3664-25G
APOSOR3664-100G
3-Methoxyphenylboronic acid 97%
3-Methoxyphenylboronic acid
Vzorec:
C₇H₉BO₃ MW: 151,96 g/mol Bod tání: 160…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161359 CAS číslo: 10365-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...