2-Methoxyphenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methoxybenzeneboronic acid, (2-Methoxyphenyl)metaboric acid, Dihydroxy(2-methoxyphenyl)borane
OR3663-100G
OR3663-25G
OR3663-5G
APOSOR3663-100GEA
1990
CZK
APOSOR3663-100G
APOSOR3663-25G
APOSOR3663-5G
2-Methoxyphenylboronic acid
2-Methoxyphenylboronic acid
Vzorec:
C₇H₉BO₃ MW: 151,96 g/mol Bod tání: 102…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00236047 CAS číslo: 5720-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...