Tisk…

7-Bromo-2-chloroquinoline 97%

Dodavatel: Apollo Scientific
Varování

OR3654-5G OR3654-25G
APOSOR3654-5GEA 1910 CZK
APOSOR3654-5G APOSOR3654-25G
7-Bromo-2-chloroquinoline 97%
7-Bromo-2-chloroquinoline
Vzorec: C₉H₅BrClN
MW: 242.5000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738671
CAS číslo: 99455-15-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR