N(α)-Boc-L-histidine
Dodavatel: Apollo Scientific
Synonyms:
Boc-His-OH
OR36354-100G
APOSOR36354-100GEA
2900
CZK
APOSOR36354-100G
N(α)-Boc-L-histidine
N(α)-Boc-L-histidine
Vzorec:
C₁₁H₁₇N₃O₄ MW: 255,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065576 CAS číslo: 17791-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...