4-Chloro-2,6-dimethylquinoline
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR3632-100MG
OR3632-250MG
APOSOR3632-100MGEA
622
CZK
APOSOR3632-100MG
APOSOR3632-250MG
4-Chloro-2,6-dimethylquinoline
4-Chloro-2,6-dimethylquinoline
Vzorec:
C₁₁H₁₀ClN Storage Temperature: Ambient |
MDL Number:
MFCD00272394 CAS číslo: 6270-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...