Tisk…

2,4,6-Trimethoxyaniline

Dodavatel: Apollo Scientific
Varování

OR3630-1G OR3630-250MG
APOSOR3630-1GEA 7940 CZK
APOSOR3630-1G APOSOR3630-250MG
2,4,6-Trimethoxyaniline
2,4,6-Trimethoxyaniline
Vzorec: C₉H₁₃NO₃
MW: 183,21 g/mol
Bod varu: 125…127 °C/0,1mmHg
Storage Temperature: Ambient
MDL Number: MFCD00017165
CAS číslo: 14227-17-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR