5-Bromoanthranilic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-5-bromobenzoic acid
OR3626-100G
OR3626-25G
APOSOR3626-100GEA
992
CZK
APOSOR3626-100G
APOSOR3626-25G
5-Bromoanthranilic acid 98%
5-Bromoanthranilic acid
Vzorec:
C₇H₆BrNO₂ Bod tání: 213…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007823 CAS číslo: 5794-88-7 EINECS: 227-338-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...