3,4,5-Trichlorophenylboronic acid pinacol ester 96%
Dodavatel: Apollo Scientific
Synonyms:
3,4,5-Trichlorophenylboronic acid,pinacol ester, 4,4,5,5-Tetramethyl-2-(3,4,5-trichlorophenyl)-1,3,2-dioxaborolane
OR361673-1G
OR361673-5G
APOSOR361673-1GEA
2650
CZK
APOSOR361673-1G
APOSOR361673-5G
3,4,5-Trichlorophenylboronic acid pinacol ester 96%
3,4,5-Trichlorophenylboronic acid pinacol ester
Vzorec:
C₁₂H₁₄BCl₃O₂ MW: 307,41 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11504973 CAS číslo: 942069-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...