1-(Toluene-4-sulphonyl)indole-3-boronic acid pinacol ester
Dodavatel: Apollo Scientific
Synonyms:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-indole
OR361656-1G
OR361656-25G
OR361656-5G
APOSOR361656-1GEA
4300
CZK
APOSOR361656-1G
APOSOR361656-25G
APOSOR361656-5G
1-(Toluene-4-sulphonyl)indole-3-boronic acid pinacol ester
1-(Toluene-4-sulphonyl)indole-3-boronic acid pinacol ester
Vzorec:
C₂₁H₂₄BNO₄S MW: 397,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08063118 CAS číslo: 1073354-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...