Tisk…

Tetrahydropyran-4-boronic acid pinacol ester 95+%

Dodavatel: Apollo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-(tetrahydro-2H-pyran-4-yl)-1,3,2-dioxaborolane

OR361629-1G OR361629-5G
APOSOR361629-1GEA 2780 CZK
APOSOR361629-1G APOSOR361629-5G
Tetrahydropyran-4-boronic acid pinacol ester 95+%
Tetrahydropyran-4-boronic acid pinacol ester
Vzorec: C₁₁H₂₁BO₃
MW: 212.0936 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11506065
CAS číslo: 1131912-76-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR