Tisk…

4-(1-Piperazinylcarbonyl)phenylboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific

OR361540-1G OR361540-5G
APOSOR361540-1GEA 2280 CZK
APOSOR361540-1G APOSOR361540-5G
4-(1-Piperazinylcarbonyl)phenylboronic acid pinacol ester 98%
4-(1-Piperazinylcarbonyl)phenylboronic acid pinacol ester
Vzorec: C₁₇H₂₅BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06657704
CAS číslo: 864754-07-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR