3-Methyl-4-pyridineboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Picoline-4-boronic acid, (3-Methylpyridin-4-yl)boronic acid
OR361538-1G
OR361538-5G
APOSOR361538-1GEA
4300
CZK
APOSOR361538-1G
APOSOR361538-5G
3-Methyl-4-pyridineboronic acid 98%
3-Methyl-4-pyridineboronic acid
Vzorec:
C₆H₈BNO₂ MW: 136.9400 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03095365 CAS číslo: 894808-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...