Tisk…

2-Morpholino-3-pyridineboronic acid 98%

Dodavatel: Apollo Scientific

OR361503-1G OR361503-5G
APOSOR361503-1GEA 7410 CZK
APOSOR361503-1G APOSOR361503-5G
2-Morpholino-3-pyridineboronic acid 98%
2-Morpholino-3-pyridineboronic acid
Vzorec: C₉H₁₃BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11867876
CAS číslo: 1218790-86-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR