1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4,4,5,5-tetramethyl-2-[1-(methylethyl)pyrazol-4-yl]-1,3,2-dioxaborolane, 1-(1-Methylethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isopropyl-1H-pyrazol-4-boronic acid, pinacol ester, 1-Isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR361314-5G
APOSOR361314-5GEA
1820
CZK
APOSOR361314-5G
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester 98%
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester
Vzorec:
C₁₂H₂₁BN₂O₂ MW: 236,12 g/mol Bod tání: 36…44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11867881 CAS číslo: 879487-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...