Tisk…

2,4-Dichloro-5-methoxyphenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2,4-Dichloro-5-methoxyphenyl)boronic acid

OR361110-1G
APOSOR361110-1GEA 5130 CZK
APOSOR361110-1G
2,4-Dichloro-5-methoxyphenylboronic acid 98%
2,4-Dichloro-5-methoxyphenylboronic acid
Vzorec: C₇H₇BCl₂O₃
MW: 220,85 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12546454
CAS číslo: 431942-67-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR